Learn More
Technical Buffer Solution pH 4.01, WTW™
GREENER_CHOICE
Product Code. 10256254
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Product Code. 10256254
Brand: WTW 108811
Technical buffering solutions for pH 2.00, pH 4.01, pH 7.00 and pH 10.01: pH 4.01, pH 7.00. This allows to adapt the bulk sizes easily to individual use. The buffer 10.0 is a special case:It is only still suitable for inoLab™ device of the series level 1, 2, 3, portable meters 3xx as well as for various other old devices. All modern technical buffering solutions are traceable to PTB (Physical Technical Government Agency) or NIST (National Institute of Standards and Technology, USA). Their accuracy is ± 0,03 pH.
TRUSTED_SUSTAINABILITY
Specifications
pH 4.01
Quantity 250 mL
Packaging 25 Bottles