Learn More
Techne™ 5Prime Thermal Cyclers: Prime Gradient Upgrade
This is an UPGRADE only. Upgrade the base model of Techne 5Prime Thermal Cycler to gradient capability.
Brand:  Techne™ 93945-57
Code : 50
Additional Details : Weight : 0.01000kg
Product Code. 12612956
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Prime units can be upgraded to achieve gradient capability. The gradient function can be enabled at any time without the need to change a block. The simple process is installed via an unlock code.
Specifications
Specifications
Prime Gradient Upgrade