missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tecalcet Hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Tecalcet Hydrochloride (R 568 Hydrochloride), an orally active calcimimetic compound, allosterically and positively modulates the calcium-sensing receptor (CaSR). Tecalcet Hydrochloride (R 568 Hydrochloride) increases the sensitivity to activation by extracellular Ca2+.
Specifications
Specifications
| Chemical Name or Material | Tecalcet Hydrochloride |
| CAS | 177172-49-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H23Cl2NO |
| Quantity | 10 mg |
| Synonym | R-568 hydrochloride |
| Solubility Information | DMSO : 50 mg/mL (146.93 mM; Need ultrasonic) |
| SMILES | C[C@@H](NCCCC1=CC=CC=C1Cl)C2=CC=CC(OC)=C2.[H]Cl |
| Molecular Weight (g/mol) | 340.29 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction