missing translation for 'onlineSavingsMsg'
Learn More

Tebuconazole, MedChemExpress

Product Code. p-200097438
Encompass_Preferred
Click to view available options
Quantity:
200 mg
1 g
Unit Size:
1g
200mg
This item is not returnable. View return policy

Product Code. 16451446

Brand: Medchem Express HYB0852

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tebuconazole is an agricultural azole fungicide which can also inhibit CYP51 with IC50s of 0.9 and 1.3 μM for Candida albicans CYP51 (CaCYP51) and truncated Homo sapiens CYP51 (δ60HsCYP51), respectively.

Tebuconazole is an agricultural azole fungicide which can also inhibit CYP51 with IC50s of 0.9 and 1.3 μM for Candida albicans CYP51 (CaCYP51) and truncated Homo sapiens CYP51 (δ60HsCYP51), respectively.

Specifications

Chemical Name or Material Tebuconazole
Physical Form Solid
CAS 107534-96-3
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C16H22ClN3O
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 50 mg/mL (162.43 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC(C)(C)C(O)(CCC1=CC=C(Cl)C=C1)CN2C=NC=N2
Molecular Weight (g/mol) 307.82
Formula Weight 307.82
Percent Purity 99.64%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.