missing translation for 'onlineSavingsMsg'
Learn More

Tebuconazole 100 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30322321
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322321 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322321 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010371ME
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tebuconazole
CAS 107534-96-3
Analyte or Component Names Tebuconazole
Shipping Condition Room Temperature
Molecular Formula C16H22ClN3O
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 1H-1,2,4-Triazole-1-ethanol, α-[2-(4-chlorophenyl)ethyl]-α-(1,1-dimethylethyl)-, (±)-, α-[2-(4-Chlorophenyl)ethyl]-α-(1,1-dimethylethyl)-1H-1,2,4-triazole-1-ethanol, (±)-Tebuconazole, Atlas, BAY-HWG 1608, Corail, Dedalus 25 WDG, Elite, Elite (fungicide), Ethyltrianol, Etiltrianol, Fenetrazole, Folicur, Folicur 200EC, Folicur 3.6F, Gelseal, Gelseal (fungicide), Greenseal, HWG 1608, Hao Li-ke, Horizon, Horizon (fungicide), Horizont, Microban S 2142, Microban TZ, ONLY ONE Flowable, Orius, Orius (fungicide), Orius 25EC, Preventol A 8, Raxil, Raxil 2DS, Raxil Ultra, Rival 200EC, Riza, Rosacur, S 2142, Silvacure, Sparta, Sparta 250EW, Tebu 250EW, Tebu 60, Tebuconazole, Terbutrazole, Timbertreat T;Formerly 80443-41-0
InChI Formula InChI=1S/C16H22ClN3O/c1-15(2,3)16(21,10-20-12-18-11-19-20)9-8-13-4-6-14(17)7-5-13/h4-7,11-12,21H,8-10H2,1-3H3
SMILES CC(C)(C)C(O)(CCc1ccc(Cl)cc1)Cn2cncn2
IUPAC Name 1-(4-chlorophenyl)-4,4-dimethyl-3-(1,2,4-triazol-1-ylmethyl)pentan-3-ol
Solvent or Matrix Methanol
Molecular Weight (g/mol) 307.82
Quantity 1 mL
Formula Weight 307.1451 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.