missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tebentafusp Humanized Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA554999

Product Code. 30132108

  • £441.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Compatible with Direct ELISA.

Tebentafusp targets gp100 peptide presented by HLA-A*02:01 on melanoma cells and redirects T cells to kill these cancer cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tebentafusp Humanized
Recombinant Monoclonal
Unconjugated
CD3E; T3E
Recombinant Human CD3E and TCR (target: gp100 peptide-HLA Complex).
100 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA
1.06 mg/mL
9mM citric acid, 41mM sodium phosphate with 2mM DTT, 1% mannitol, 5% trehalose, 0.02% Tween 20 and no preservative; pH 6.5
Human
Metal-chelate chromatography
RUO
Human
Antibody
scFv
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.