missing translation for 'onlineSavingsMsg'
Learn More

Tebbe reagent, 0.5M solution in toluene

Código de producto. 10089001 Tienda Thermo Scientific Chemicals Productos
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
AcroSeal™ Glass bottle
Glass Bottle
Tamaño de la unidad:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Packaging unitSize
10089001 100 mL AcroSeal™ Glass bottle 100mL
10772263 25 mL Glass Bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10089001 Proveedor Thermo Scientific Acros N.º de proveedor 368971000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 67719-69-1 | C13H18AlClTi

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination), Lewis acid
TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Tebbe reagent
Name Note 0.5M solution in toluene
CAS 67719-69-1, 108-88-3
Color Purple to Red
Density 0.9270g/mL
Flash Point 4°C
Packaging AcroSeal™ Glass bottle
Molecular Formula C13H18AlClTi
MDL Number MFCD00151575
Quantity 100 mL
Fieser 08,83; 10,87; 11,52; 12,54; 13,71; 15,79; 16,70; 17,69
Specific Gravity 0.927
Synonym Bis(cyclopentadienyl-μ-chloro(dimethylaluminum-μ-methylenetitanium
Solubility Information Solubility in water: reacts.
Formula Weight 284.62
Physical Form Solution
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Tebbe reagent, 0.5M solution in toluene
Palabra de advertencia
  • Danger
Clasificación
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.