missing translation for 'onlineSavingsMsg'
Learn More

Tebbe reagent, 0.5 M solution in toluene, Thermo Scientific™

Product Code. 17535535 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity: 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17535535 Quantity: 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17535535 Supplier Thermo Scientific Acros Supplier No. 368970050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Alpha Vector TEBBEREAGENT
CAS 67719-69-1, 108-88-3
CAS Min % 15%, 85%
CAS Max % 15%, 85%
Color Dark red to purple
Flash Point 4°C
Packaging Glass Bottle
Molecular Formula C13H18AlClTi
MDL Number 00151575
Quantity 5 mL
Fieser 17-69
Merck Index 15-9228
Solubility Information Reacts with water
InChI Key HIVIAXIRSNAVAD-UHFFFAOYSA-M
SMILES [Cl-].[Ti+4].C[Al](C)[CH2-].[CH-]1C=CC=C1.[CH-]1C=CC=C1
IUPAC Name titanium(4+) (dimethylalumanyl)methanide bis(cyclopenta-2,4-dien-1-ide) chloride
Molecular Weight (g/mol) 284.59
Physical Form Solution
Chemical Name or Material Tebbe reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.