missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Teashirt homolog 1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82038UV
This item is not returnable.
View return policy
Description
Teashirt homolog 1 Polyclonal antibody specifically detects Teashirt homolog 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Teashirt homolog 1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 10194 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 350 | |
| Antigen NY-CO-33, NY-CO-33, SDCCAG33, Serologically defined colon cancer antigen 33teashirt family zinc finger 1, teashirt homolog 1, teashirt zinc finger homeobox 1, TSH1teashirt zinc finger 1 | |
| Teashirt homolog 1 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human Teashirt homolog 1. The immunogen is located within amino acids 70 - 120 of Teashirt homolog 1. Amino Acid Squence: AHFKGSSSREEKEDPQ | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction