missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Tear-Away Aluminum Seals for Serum Bottles, Small

Product Code. 15113075 Shop All Labconco Corporation Products
Change view
Click to view available options
Thread Size (Metric):
57 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) unitSize
15113075 57 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15113075 Supplier Labconco™ Supplier No. 7577010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Labconco™ Tear-Away Aluminum Seals for Serum Bottles, Small are packs of 13mm caps that fit in 2mL and 3mL glass serum bottles. 100/Pack

  • Serum Bottles hold small samples for batch drying on a tray
  • They may also be connected to valve ports on drying chambers and manifolds
  • Aluminum seal caps sold separately for easy sealing

Specifications

For Use With (Application) Drying small samples
Material Aluminum
No. per Pack 100Pack
Thread Size (Metric) 57 mm
Neck Finish 57 mm
Shape Round
Product Type Aluminum Seal
For Use With (Equipment) 2 mL/3 mL Glass Serum Bottles
Closure Size 13 mm
Diameter (Metric) 13 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.