missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEAD4 Recombinant Rabbit Monoclonal Antibody (8R0N1)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555568
This item is not returnable.
View return policy
Description
Immunogen Sequence: SATAFHSSMA LARGPGRPAV SGFWQGALPG QAGTSHDVKP FSQQTYAVQP PLPLPGFESP AGPAPSPSAP PAPPWQGRSV ASSKLWMLEF SAFLEQQQDP D.
This gene product is a member of the transcriptional enhancer factor (TEF) family of transcription factors, which contain the TEA/ATTS DNA-binding domain. It is preferentially expressed in the skeletal muscle, and binds to the M-CAT regulatory element found in promoters of muscle-specific genes to direct their gene expression. Alternatively spliced transcripts encoding distinct isoforms, some of which are translated through the use of a non-AUG (UUG) initiation codon, have been described for this gene.
Specifications
| TEAD4 | |
| Recombinant Monoclonal | |
| 1.07 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q15561 | |
| Tead4 | |
| A synthetic peptide corresponding to a sequence within amino acids 140-240 of human TEAD4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 8R0N1 | |
| Unconjugated | |
| Tead4 | |
| EFTR-2; Etfr2; ETFR-2; ETF-related factor 2; ETF-related factor-2; FGF-regulated 19; FR-19; hRTEF-1B; related transcription enhancer factor 1B; RTEF1; RTEF-1; TCF13L1; Tcf13r1; TEA domain family member 4; TEA domain transcription factor 4; TEAD4; TEAD-4; TEF-1-related factor 1; TEF-1-related factor FR-19; TEF3; TEF-3; Tefr; Tefr1; TEFR-1; Tefr1a; Transcription factor 13-like 1; transcription factor RTEF-1; transcriptional enhancer factor 1-related; transcriptional enhancer factor 3; transcriptional enhancer factor TEF-3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7004 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction