missing translation for 'onlineSavingsMsg'
Learn More

TEAD4 Antibody (5H3), Novus Biologicals™

Product Code. 18392139 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392139 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392139 Supplier Novus Biologicals Supplier No. H00007004M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TEAD4 Monoclonal antibody specifically detects TEAD4 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEAD4
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, KnockDown
Classification Monoclonal
Clone 5H3
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_003204
Gene Alias EFTR-2, hRTEF-1B, related transcription enhancer factor 1B, RTEF1RTEF-1, TCF13L1MGC9014, TEA domain family member 4TEF3, TEAD-4, TEF-3, TEFR-1, Transcription factor 13-like 1, Transcription factor RTEF-1, transcriptional enhancer factor 1-related, transcriptional enhancer factor 3, transcriptional enhancer factor TEF-3
Host Species Mouse
Immunogen TEAD4 (NP_003204, 151 a.a. ~ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ARGPGRPAVSGFWQGALPGQAGTSHDVKPFSQQTYAVQPPLPLPGFESPAGPAPSPSAPPAPPWQGRSVASSKLWMLEFSAFLEQQQDPDTYNKHLFVHIGQSSPSYSDP
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7004
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.