missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEAD3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£176.00 - £393.00
Specifications
| Antigen | TEAD3 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18643991
|
Novus Biologicals
NBP2-93128-0.02ml |
0.02 mL |
£176.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18605320
|
Novus Biologicals
NBP2-93128-0.1ml |
0.1 mL |
£393.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TEAD3 Polyclonal antibody specifically detects TEAD3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| TEAD3 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 7005 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DTEF-1, ETFR-1, TEA domain family member 3TEAD-3, TEAD5TEF5TEA domain family member 5, TEF-5, transcriptional enhancer factor 5, transcriptional enhancer factor TEF-5, Transcriptional enhancer factor TEF-5 (DTEF-1) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-330 of human TEAD3 (NP_003205.2). SSAQIVSASVLQNKFSPPSPLPQAVFSTSSRFWSSPPLLGQQPGPSQDIKPFAQPAYPIQPPLPPTLSSYEPLAPLPSAAASVPVWQDRTIASSRLRLLEYSAFMEVQRDPDTYSKHLFVHIGQTNPAFSDPPLEAVDVRQIYDKFPEKKGGLKELYEKGPPNAFFLVKFWADLNSTIQEGPGAFYGVSSQYSSADSMTIS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title