missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEAD2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TEAD2 |
|---|---|
| Concentration | 0.29 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16810775
|
Proteintech
21159-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16800775
|
Proteintech
21159-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TEAD2 is a putative transcription factor that binds to the SPH and GT-IIC enhansons (5′-GTGGAATGT-3′). TEAD2 may be involved in the gene regulation of neural development. TEAD2 binds to the M-CAT motif.Specifications
| TEAD2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| P48301, Q15562 | |
| 21677, 8463 | |
| TEAD2 Fusion Protein Ag14074 | |
| Primary | |
| -20°C | |
| TEAD2 |
| 0.29 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| ETF, TEA domain family member 2, TEAD 2, TEAD2, TEF 4, TEF4 | |
| TEAD2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title