missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEAD1/2/3/4 Recombinant Rabbit Monoclonal Antibody (23GB4680), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552728
This item is not returnable.
View return policy
Description
TEAD1, TEAD2, TEAD3, and TEAD4 are members of the TEAD (TEA domain) family of transcription factors. These factors are known to play important roles in various biological processes, including development, cell proliferation, differentiation, and organogenesis. TEAD1, TEAD2, TEAD3, and TEAD4 exhibit overlapping but also distinct expression patterns and functions. They are involved in diverse cellular processes and have been implicated in the regulation of embryonic development, tissue homeostasis, cell fate determination, and tumor progression. TEAD proteins are known to interact with various co-factors, including the transcriptional co-activators YAP (Yes-associated protein) and TAZ (transcriptional co-activator with PDZ-binding motif), which play crucial roles in the Hippo signaling pathway. The interaction between TEAD and YAP/TAZ is essential for the activation of downstream target genes involved in cell proliferation and survival.
Specifications
| TEAD1/2/3/4 | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P28347 | |
| TEAD1 | |
| A synthesized peptide derived from human TEAD1+TEAD2+TEAD3+TEAD4 (150-300AA). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4680 | |
| Unconjugated | |
| TEAD1 | |
| DTEF-1; ETF; ETFR-1; Protein GT-IIC; RTEF1; RTEF-1; TCF13; TCF-13; TEA Domain Family Member 1; TEA Domain Family Member 1 (SV40 Transcriptional Enhancer Factor); TEA Domain Family Member 2; TEA Domain Family Member 3; TEA Domain Family Member 5; TEA Domain Transcription Factor 1; TEA Domain Transcription Factor 2; TEA Domain Transcription Factor 3; TEAD; TEAD2; TEAD-2; TEAD3; TEAD-3; TEAD-4; TEAD5; TEF3; TEF4; TEF-4; TEF5; TEF-5; Transcription Factor 13; Transcriptional Enhancer Factor 1; Transcriptional Enhancer Factor 1-Related; Transcriptional Enhancer Factor 5; Transcriptional Enhancer Factor TEF-1; Transcriptional Enhancer Factor TEF-4; Transcriptional Enhancer Factor TEF-5; Transcriptional Enhancer Factor TEF-5 (DTEF-1) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 361630, 7003 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction