missing translation for 'onlineSavingsMsg'
Learn More

TE Buffer, Tris-EDTA, 10X Solution, pH 7.6, Molecular Biology, Fisher BioReagents™

Product Code. 10409123
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Unit Size:
1 L
100mL
500mL
This item is not returnable. View return policy

Product Code. 10409123

Brand: Fisher BioReagents BP24751

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

TE Buffer, Tris-EDTA, 10X Solution, pH 7.6, Molecular Biology

  • Save time and simplify your buffer preparation step by using Fisher BioReagents 10X TE buffer solution - simply dilute as needed
  • 10X solution contains 100mM tris and 10mM EDTA
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TE solution with 9 volumes of ultrapure water, such as BP2819
  • Filtered through a 5 micron filter and autoclaved
  • Tested for the absence of DNase, RNase, and protease
  • TE buffer is routinely used for resuspending nucleic acid samples
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris-EDTA (TE), 10X Solution
Name Note pH 7.6
CAS 77-86-1, 6381-92-6, 7647-01-0, 7732-18-5
Color Colorless
pH 7.4 to 8.1
Physical Form Liquid
Quantity 1 L
DNase DNase free
Filtered Through Filtered through a 5-micron filter and autoclaved.
Packaging Nalgene Poly Bottle
Protease Protease free
Synonym TE
Grade Molecular Biology
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.