Learn More
TE buffer, pH 8.0, RNAse free, Thermo Scientific Chemicals
£85.30 - £87.00
Chemical Identifiers
CAS | 1185-53-1 |
---|---|
Molecular Formula | C4H12ClNO3 |
Molecular Weight (g/mol) | 157.594 |
MDL Number | MFCD00236359 |
InChI Key | QKNYBSVHEMOAJP-UHFFFAOYSA-N |
PubChem CID | 93573 |
IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
SMILES | C(C(CO)(CO)N)O.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15445059
|
Thermo Scientific Alfa Aesar
J62745.=E |
125 mL |
£87.00
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15455059
|
Thermo Scientific Alfa Aesar
J62745.AK |
250 mL |
£85.30
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Trizma is used in the formulation of buffer solutions in the pH range between 7.5 and 8.5. Tris buffer solutions are widely used in cell and molecular biology for processes such as protein and nucleic acid extraction and purification. Tris-EDTA (TE) buffer solution, pH 8.0 may also be used as a washing buffer. TE buffer is often used to store DNA and RNA while protecting it from degradation. Some protocols use TE 10:0.1 with 0.1 mM EDTA to reduce the interaction of the EDTA with downstream applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTrizma is used in the formulation of buffer solutions in the pH range between 7.5 and 8.5. Tris buffer solutions are widely used in cell and molecular biology for processes such as protein and nucleic acid extraction and purification. Tris-EDTA (TE) buffer solution, pH 8.0 may also be used as a washing buffer. TE buffer is often used to store DNA and RNA while protecting it from degradation. Some protocols use TE 10:0.1 with 0.1 mM EDTA to reduce the interaction of the EDTA with downstream applications.
Solubility
Not miscible or difficult to mix in water.
Notes
200mM Tris-HCl with 20mM EDTA
Chemical Identifiers
1185-53-1 | |
157.594 | |
QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
C4H12ClNO3 | |
MFCD00236359 | |
93573 | |
C(C(CO)(CO)N)O.Cl |
Specifications
1185-53-1 | |
Liquid | |
MFCD00236359 | |
QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride | |
93573 |
8.0 | |
C4H12ClNO3 | |
Not miscible or difficult to mix in water. | |
C(C(CO)(CO)N)O.Cl | |
157.594 | |
TE buffer, RNAse free |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only