missing translation for 'onlineSavingsMsg'
Learn More

TE Buffer, 50X Solution, Molecular Biology Grade, Ultrapure, Thermo Scientific™

Product Code. 15805418 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500mL
Packaging:
Nalgene bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15805418 500mL Nalgene bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15805418 Supplier Thermo Scientific Supplier No. J75834.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1185-53-1
Quantity 500mL
Molecular Formula C4H12ClNO3
Packaging Nalgene bottle
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
SMILES C(C(CO)(CO)N)O.Cl
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
Molecular Weight (g/mol) 157.594
PubChem CID 93573
Grade Molecular Biology
Chemical Name or Material TE Buffer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.