missing translation for 'onlineSavingsMsg'
Learn More

Cytiva TE 62 Transfer Unit Accessory: Gel Cassette

Product Code. 12383929 Shop All Cytiva Products
Click to view available options
Type:
Gel Cassette
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12383929

Brand: Cytiva 80620654

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

With two foam sponges 3mm thick and 1 foam sponge 6mm thick

Replacement gel cassette for use with Cytiva TE 62 Transfer Unit

  • Hinged cassettes, molded of tough, glass-filled polyethersulfone for high durability, provide equal pressure across the gel
  • Cassettes load easily, with even contact between the gel and transfer membrane

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) TE 62 Protein Transfer Unit
Type Gel Cassette
Includes Two Foam sponges 3mm thick and 1 Foam sponge 6mm thick
Quantity Complete packsize
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.