missing translation for 'onlineSavingsMsg'
Learn More

Cytiva TE 62 Transfer Unit Accessory: Electrode Panel Assembly

Product Code. 12941640 Shop All Cytiva Products
Change view
Click to view available options
:
Electrode panel; Color: black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12941640 Electrode panel; Color: black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12941640 Supplier Cytiva Supplier No. 80620616

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

  • Safe, sturdy electrode panels lock automatically in place
  • Choose from black or gray panels

Two panels are required to run the unit.

TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) TE 62 Protein Transfer Unit
Type Electrode Panel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.