missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ TDZD 8

Product Code. 18781799
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18781799 50 mg 50mg
18719508 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18781799 Supplier R&D Systems™ Supplier No. 6092/50
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective non-ATP competitive inhibitor of GSK 3β

TDZD 8 is a selective non-ATP competitive inhibitor of GSK 3β (IC50 = 2 μM); thiadiazolidinone deriviative. Does not inhibit Cdk-1/cyclin B, CK-II, PKA or PKC at >100 μM. Reduces severity of L-dopa-induced dyskinesia in a Parkinson's disease in vivo model.
TRUSTED_SUSTAINABILITY

Specifications

CAS 327036-89-5
Quantity 50 mg
Solubility Soluble to 100mM in DMSO, 100mM in ethanol
Product Line Tocris
Molecular Formula C10H10N2O2S
Formula Weight 222.26g/mol
Color White
Product Type Inhibitor

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.