missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific TDX QS7-PRO 384W QPCR,DESKTOP EACH
Brand: Thermo Scientific A43185TDX
52532.20 GBP valid until 2026-05-31
BEST PRICE on Promo! Use promo code "27998" to get your promotional price.
TDX QS7-PRO 384W QPCR,DESKTOP EACH
This item is not returnable.
View return policy
Description
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?