missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TdT Rabbit anti-Human, Clone: DNTT/4506R, Novus Biologicals™
Rabbit Monoclonal Antibody
£412.00
Specifications
| Antigen | TdT |
|---|---|
| Clone | DNTT/4506R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
TdT Monoclonal specifically detects TdT in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| TdT | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cell Cycle and Replication, Immunology | |
| 10 mM PBS with 0.05% BSA | |
| 1791 | |
| IgG κ | |
| Protein A or G purified |
| DNTT/4506R | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| deoxynucleotidyltransferase, terminal, EC 2.7.7, EC 2.7.7.31, nucleosidetriphosphate:DNA deoxynucleotidylexotransferase, TDTDNA nucleotidylexotransferase, Terminal addition enzyme, Terminal deoxynucleotidyltransferase, terminal deoxyribonucleotidyltransferase, Terminal transferase | |
| Synthetic peptide corresponding to residues within aa1-100 of TdT was used as an immunogen. | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title