missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TdT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5149508
This item is not returnable.
View return policy
Description
TdT Polyclonal Antibody for Western Blot, IHC (P)
Terminal Deoxynucleotidyl Transferase (TdT) is a DNA polymerase located in the cell nucleus which catalyses the polymerization of deoxynucleotides at the 3' hydroxyl ends of oligo or polydeoxynucleotide initiators and functions without a template. TdT is considered to be a highly specific marker for the diagnosis and classification of acute lymphoblastic lymphoma/leuksemias. The determination of TdT expression is most valuable when it is different to differentiate histologically between lymphoblastic lymphoma and Burkitt's lymphoma.
Specifications
| TdT | |
| Polyclonal | |
| Unconjugated | |
| DNTT | |
| Deoxynucleotidyltransferase terminal; deoxynucleotidyltransferase, terminal; DNA nucleotidylexotransferase; Dntt; Nucleosidetriphosphate DNA Deoxynucleotidylexotransferase; nucleosidetriphosphate:DNA deoxynucleotidylexotransferase; TDT; Terminal addition enzyme; terminal deoxynucleotidyl transferase; terminal deoxynucleotidyltransferase; terminal deoxyribonucleotidyltransferase; terminal transferase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1791 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P04053 | |
| DNTT | |
| Partial recombinant protein made to an internal sequence of human TdT (between residues 50-300). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction