Learn More
TdT Monoclonal Antibody (7UNA8U6), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-9739-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The monoclonal antibody 7UNA8U6 reacts with human, mouse, and rat TdT. TdT, or terminal-deoxynucleotidyl transferase, is a 60 kDa polymerase responsible for the template-independent addition of N-nucleotides at gene segment junctions of developing lymphocytes. Expression is found in the nucleus of immature lymphocytes but not in mature lymphocytes or non-lymphoid cells. TdT plays a critical role during TCR and Ig gene rearrangement and functions to increase the diversity of the lymphocyte repertoire through the addition of N-nucleotides onto the ends of newly formed P-nucleotides. After positive selection, expression of TdT becomes decreased or absent. Expression of TdT has been shown to be present in lymphoma or lymphoblastic leukemia cells. Applications Reported: This 7UNA8U6 antibody has been reported for use in western blotting and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections. Applications Tested: This 7UNA8U6 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded human and mouse tissue using low pH antigen retrieval and can be used at less than or equal to 10 μg/mL. This 7UNA8U6 antibody has also been tested by western blot analysis of reduced cell lysate prepared from mouse and rat cells and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Terminal Deoxynucleotidyl Transferase (TdT) is a DNA polymerase located in the cell nucleus which catalyses the polymerization of deoxynucleotides at the 3' hydroxyl ends of oligo or polydeoxynucleotide initiators and functions without a template. TdT is considered to be a highly specific marker for the diagnosis and classification of acute lymphoblastic lymphoma/leuksemias. The determination of TdT expression is most valuable when it is different to differentiate histologically between lymphoblastic lymphoma and Burkitt's lymphoma.Specifications
TdT | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P04053, P09838 | |
DNTT | |
Affinity chromatography | |
RUO | |
1791, 21673, 294051 | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
7UNA8U6 | |
Unconjugated | |
DNTT | |
Deoxynucleotidyltransferase terminal; deoxynucleotidyltransferase, terminal; DNA nucleotidylexotransferase; Dntt; Nucleosidetriphosphate DNA Deoxynucleotidylexotransferase; nucleosidetriphosphate:DNA deoxynucleotidylexotransferase; TDT; Terminal addition enzyme; terminal deoxynucleotidyl transferase; terminal deoxynucleotidyltransferase; terminal deoxyribonucleotidyltransferase; terminal transferase | |
Mouse | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 κ |
For Research Use Only.