missing translation for 'onlineSavingsMsg'
Learn More

TDS Standard Solutions 800mg/L (ppm), Hanna Instruments™

Product Code. 11514307
Change view
Click to view available options
Quantity:
25 x 20 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11514307 25 x 20 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11514307 Supplier Hanna Instruments Supplier No. HI70080P
  • ONLINE EXCLUSIVE PRICE
    £63.69
    £52.18 / Pack of 1
    Save £11.51
    18% Off
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Packaging Sachets
Odor Odorless
Physical Form Liquid
Color Colorless
pH 7
Solubility Soluble
Quantity 25 x 20 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.