missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDS Standard Solution, 1382ppm, Reagecon™
Click to view available options
Quantity:
500 mL
Description
Fisher Scientific offer a comprehensive range of conductivity standards, traceable to NIST reference materials. The range includes a standard range as well as a broader range of very low and high values, offering increased flexibility.
- Accurate to ±1%
- Traceable to NIST
- GLP compliant
- Certificate of analysis supplied with each bottle (including method of analysis and standard reference material detail)
- Wide range: 20 values from 1.3μS/cm to 500,000μS/cm inclusive
- An enhanced shelf life is assured through the use of proprietary technologyAccurate conductivity measurement requires periodic calibration of the conductivity sensor in order to determine its cell constant (the multiplier used to convert the sensor's output to the displayed conductivity). Our standard value conductivity standards match the pre-programmed values used by all leading brands of conductivity meters for automatic calibration, While the broad range standards allow flexibility.
Specifications
Concentration | 1382ppm |
Quantity | 500 mL |
Certificate of Analysis | Yes |
Methods | Standard solution verified by Reference Materials Laboratory |
Solvent or Matrix | Aqueous |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'