missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDS Calibration Solution 1500 mg/L (ppm), Hanna Instruments™
Click to view available options
Quantity:
25 x 20 mL
Unit Size:
Pack of 25
Specifications
Specifications
| Packaging | Sachets |
| Odor | Odorless |
| Physical Form | Liquid |
| Color | Colorless |
| pH | 7 |
| Solubility | Soluble |
| Quantity | 25 x 20 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction