missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TDRKH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100348
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TDRKH.
TDRKH is a protein of unknown function containing one tudor domain, and two KH domains. It has been found to interact with PIWIL1 and PIWIL4 by binding to methylated arginines residues.
Specifications
| TDRKH | |
| Polyclonal | |
| Unconjugated | |
| TDRKH | |
| 2700091C21Rik; putative RNA binding protein; RGD1311196; RP11-98D18.8; Tdrd2; TDRD7; Tdrkh; tudor and KH domain containing; tudor and KH domain containing protein; tudor and KH domain-containing protein; tudor domain containing 2; tudor domain-containing protein 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11022, 310652, 72634 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q80VL1, Q9Y2W6 | |
| TDRKH | |
| A synthesized peptide derived from human TDRKH(Accession Q9Y2W6), corresponding to amino acid residues A248-P298. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction