missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDRKH Antibody (PCRP-TDRKH-1H2) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14276JF669
This item is not returnable.
View return policy
Description
TDRKH Monoclonal antibody specifically detects TDRKH in Human samples. It is validated for Flow Cytometry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| TDRKH | |
| Monoclonal | |
| Janelia Fluor 669 | |
| TDRD2, tudor and KH domain containing, tudor and KH domain-containing protein | |
| Recombinant full-length human TDRKH protein (Uniprot: Q9Y2W6 ) | |
| 0.1 mL | |
| Cell Biology | |
| 11022 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Flow Cytometry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-TDRKH-1H2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction