missing translation for 'onlineSavingsMsg'
Learn More

TDRKH Antibody (PCRP-TDRKH-1H2), Allophycocyanin, Novus Biologicals™

Product Code. 18358013
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358013 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18358013 Supplier Bio-Techne Supplier No. NBP314276APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TDRKH Monoclonal antibody specifically detects TDRKH in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRKH
Applications Flow Cytometry
Classification Monoclonal
Clone PCRP-TDRKH-1H2
Conjugate APC
Dilution Flow Cytometry
Formulation PBS
Gene Alias TDRD2, tudor and KH domain containing, tudor and KH domain-containing protein
Host Species Mouse
Immunogen Recombinant full-length human TDRKH protein (Uniprot: Q9Y2W6 )
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 11022
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.