missing translation for 'onlineSavingsMsg'
Learn More

TDRKH Antibody, Novus Biologicals™

Product Code. 18399281 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399281 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399281 Supplier Novus Biologicals Supplier No. NBP312498

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TDRKH Polyclonal antibody specifically detects TDRKH in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRKH
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:150
Formulation Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Gene Alias TDRD2, tudor and KH domain containing, tudor and KH domain-containing protein
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 480-515 on TDRD2 protein.
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 11022
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.