missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TDRD7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15868473
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15868473 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15868473 Supplier Invitrogen™ Supplier No. PA570066

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

TDRD7, also known as Tudor domain containing protein 7, is a component of chromatoid bodies in spermatids (named processing bodies or P-bodies in somatic cells). It is found in a mRNP complex together with TDRD1, TDRD6 and DDX4. It is also found in a complex with CABLES1, PCTK2 and PIWIL1. TDRD7 has been shown to interact with histone H3 tri-methylated at K9 in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRD7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TDRD7
Gene Accession No. Q9R1R4
Gene Alias 5730495N10Rik; AI447470; CATC4; PCTAIRE2-binding protein; Pctaire2bp; RP11-508D10.1; TDRD7; TRAP; tudor domain containing 7; tudor domain-containing protein 7; tudor repeat associator with PCTAIRE 2; tudor repeat associator with PCTAIRE-2
Gene Symbols TDRD7
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human TDRD7 (aa 813-862).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85425
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.