missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TDRD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558079
This item is not returnable.
View return policy
Description
Immunogen sequence: TENNIVVDKS DLIPKVLTLN VGDEFCGVVA HIQTPEDFFC QQLQSGRKLA ELQASLSKYC DQLPPRSDFY PAIGDICCAQ Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 76%.
This gene is similar to a mouse gene that encodes a tudor domain protein. Alternatively spliced transcript variants have been described but their full length sequences have not been determined.
Specifications
| TDRD1 | |
| Polyclonal | |
| Unconjugated | |
| Tdrd1 | |
| Cancer/testis antigen 41.1; CT41.1; FLJ21082; MTR-1; Tdrd1; testicular tissue protein Li 193; tudor domain containing 1; tudor domain containing protein 1; tudor domain-containing protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56165 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BXT4 | |
| Tdrd1 | |
| Recombinant protein corresponding to Human TDRD1. Recombinant protein control fragment (Product #RP-97027). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction