missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDRD1 Monoclonal Antibody (739206), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA524351
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
TDRD1 Monoclonal antibody specifically detects TDRD1 in Mouse samples. It is validated for Immunohistochemistry (Frozen).Specifications
TDRD1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q99MV1 | |
Tdrd1 | |
E. coli-derived recombinant mouse TDRD1 Asn831-Lys960 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Immunohistochemistry (Frozen) | |
739206 | |
Unconjugated | |
Tdrd1 | |
Cancer/testis antigen 41.1; CT41.1; FLJ21082; MTR-1; Tdrd1; testicular tissue protein Li 193; tudor domain containing 1; tudor domain containing protein 1; tudor domain-containing protein 1 | |
Rat | |
Protein A/G | |
RUO | |
83561 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |