missing translation for 'onlineSavingsMsg'
Learn More

TDRD1 Antibody, Novus Biologicals™

Product Code. 18003178 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
Unit Size:
0.05mg
0.10mg
25µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18003178 0.1 mg 0.10mg
18041019 0.025 mg 25µg
18075668 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18003178 Supplier Novus Biologicals Supplier No. NBP211487

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TDRD1 Polyclonal specifically detects TDRD1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRD1
Applications Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:10000, ELISA 1:500, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:100
Formulation Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide
Gene Alias CT41.1Cancer/testis antigen 41.1, FLJ21082, tudor domain containing 1, tudor domain containing protein 1, tudor domain-containing protein 1
Gene Symbols TDRD1
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 570-605 on TDRD1 protein.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56165
Test Specificity This antibody detects multiple bands of approximate MW of 83, 94, and a minor 114kDa in TDRD1 samples. The antibody does not cross reacts with other histones or DNA binding proteins
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.