missing translation for 'onlineSavingsMsg'
Learn More

TDRD1 Antibody [HRP], Novus Biologicals™

Product Code. 30733567 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733567 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733567 Supplier Novus Biologicals Supplier No. AF7320H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

TDRD1 Polyclonal antibody specifically detects TDRD1 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRD1
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias CT41.1Cancer/testis antigen 41.1, FLJ21082, tudor domain containing 1, tudor domain containing protein 1, tudor domain-containing protein 1
Host Species Sheep
Immunogen E. coli-derived recombinant mouse TDRD1, Asn831-Lys960, Accession # Q99MV1
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56165
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.