missing translation for 'onlineSavingsMsg'
Learn More

TDP-43/TARDBP Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18316653 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18316653 20 μg 20µL
18372067 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18316653 Supplier Novus Biologicals Supplier No. NBP31695220UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TDP-43/TARDBP Polyclonal antibody specifically detects TDP-43/TARDBP in Human samples. It is validated for Western Blot, Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDP-43/TARDBP
Applications Western Blot, Dot Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Dot Blot
Formulation PBS, 50% glycerol, pH7.3
Gene Alias ALS10TAR DNA-binding protein-43, TAR DNA binding protein, TDP43, TDP-43, TDP-43TAR DNA-binding protein 43
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around S409 & S410 of human TDP-43/TARDBP (NP_031401.1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing, Epigenetics, Immunology, Mitochondrial Fusion Proteins, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 23435
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.