missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDP-43/TARDBP Antibody (3H8), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-92695FR
This item is not returnable.
View return policy
Description
TDP-43/TARDBP Monoclonal specifically detects TDP-43/TARDBP in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Free-Floating.
Specifications
| TDP-43/TARDBP | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TARDBP | |
| Full length recombinant human TDP43 expressed in and purified from E. coli, epitope in peptide HNSNRQLERSGRFGGNPGGF, amino acids 264-283 of the human sequence. | |
| 0.1 mL | |
| Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing, Epigenetics, Immunology, microRNA, Mitochondrial Fusion Proteins, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Virology Bacteria and Parasites | |
| 23435 | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Free Floating) | |
| 3H8 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Free-Floating | |
| ALS10TAR DNA-binding protein-43, TAR DNA binding protein, TDP43, TDP-43, TDP-43TAR DNA-binding protein 43 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine, Equine | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction