missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TDP-43 Monoclonal Antibody (3H8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547420
This item is not returnable.
View return policy
Description
TDP-43 Monoclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
TDP43 was first identified as a novel cellular protein that binds to HIV-1 virus TAR DNA sequence motifs and acts a transcriptional repressor to the HIV-1 LTR. Later experiments revealed that TDP43 also regulates the splicing of exon 9 of the cystic fibrosis transmembrane conductance regular (CFTR), most likely through the association with the UG repeats at the 3'e; splice site. A hyperphosphorylated, ubiquitinated, and cleaved form of TDP43 known as pathologic TDP43 is the major disease protein in ubiquitin-positive, tau-, and alpha-synuclein-negative frontotemporal dementia (FLTD-U). TDP43 is not related to TRBP1, and RNA binding protein that binds HIV-1 TAR RNA sequences.
Specifications
| TDP-43 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 5mM sodium azide | |
| Q13148, Q921F2 | |
| TARDBP | |
| Full length recombinant human TDP43 expressed in and purified from E. coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Bovine, Pig, Horse | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| 3H8 | |
| Unconjugated | |
| TARDBP | |
| 1190002A23Rik; ALS10; C85084; hypothetical protein; OTTHUMP00000002171; OTTHUMP00000002173; RCJMB04_5g9; RP4-635E18.2; TAR DNA binding protein; TAR DNA binding protein L homeolog; TAR DNA-binding protein 43; TAR DNA-binding protein-43; tardbp; tardbp.L; Tdp43; TDP-43; wu:fb77f02; wu:fc52g10; XELAEV_18035188mg | |
| Mouse | |
| Protein G | |
| RUO | |
| 100051482, 100739753, 230908, 23435, 298648, 540632 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction