missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TDP-43 Monoclonal Antibody (3H8)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA547322

 View more versions of this product

Product Code. 17825307

  • £1482.00 / 500µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TDP-43 Monoclonal Antibody for Western Blot, ICC/IF, IHC (PFA)

TDP43 was first identified as a novel cellular protein that binds to HIV-1 virus TAR DNA sequence motifs and acts a transcriptional repressor to the HIV-1 LTR. Later experiments revealed that TDP43 also regulates the splicing of exon 9 of the cystic fibrosis transmembrane conductance regular (CFTR), most likely through the association with the UG repeats at the 3'e; splice site. A hyperphosphorylated, ubiquitinated, and cleaved form of TDP43 known as pathologic TDP43 is the major disease protein in ubiquitin-positive, tau-, and alpha-synuclein-negative frontotemporal dementia (FLTD-U). TDP43 is not related to TRBP1, and RNA binding protein that binds HIV-1 TAR RNA sequences.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TDP-43
Monoclonal
1 mg/mL
PBS with 50% glycerol and 5mM sodium azide
Q13148, Q921F2
TARDBP
Full length recombinant human TDP43 expressed in and purified from E. coli.
500 μL
Primary
Human, Mouse, Rat, Bovine, Pig, Horse
Antibody
IgG1 κ
Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
3H8
Unconjugated
TARDBP
1190002A23Rik; ALS10; C85084; hypothetical protein; OTTHUMP00000002171; OTTHUMP00000002173; RCJMB04_5g9; RP4-635E18.2; TAR DNA binding protein; TAR DNA binding protein L homeolog; TAR DNA-binding protein 43; TAR DNA-binding protein-43; tardbp; tardbp.L; Tdp43; TDP-43; wu:fb77f02; wu:fc52g10; XELAEV_18035188mg
Mouse
Protein G
RUO
100051482, 100739753, 230908, 23435, 298648, 540632
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.