missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palintest™ Comparator Reagents
Click to view available options
Analytes:
Alkalinity (CaCO3)
Chlorine DPD 2 (Monochloramine)
Copper
Nitrate
Phosphate (LR)
Zinc
Unit Size:
Each
Pack of 200
Pack of 250
Description
- Tablet form for maximum convenience and simplicity.
- Colorimetric test.
Specifications
Specifications
| Analytes | Phosphate (LR) |
| Concentration | 0 to 4 mg/L |
| For Use With (Equipment) | Palintest comparator and disc |
| Packaging | 250/Pk. |
| Quantity | 250/Pk. |
| Test Method | Phosphate reaction with ammonium molybdate to form phospho-molybdic acid, which is reduced by ascorbic acid. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction