missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TDIF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571407
This item is not returnable.
View return policy
Description
TDIF2 Polyclonal Antibody for Western Blot
This gene is thought to be involved in chromatin remodeling and gene transcription. The encoded nuclear protein binds to and enhances the transcriptional activity of the estrogen receptor alpha, and also interacts with terminal deoxynucleotidyltransferase. The expression profile of this gene is a potential biomarker for chronic obstructive pulmonary disease.
Specifications
| TDIF2 | |
| Polyclonal | |
| Unconjugated | |
| DNTTIP2 | |
| 4930588M11Rik; AA408582; acidic 82 kDa protein mRNA; AU014960; deoxynucleotidyltransferase terminal interacting protein 2; deoxynucleotidyltransferase terminal-interacting protein 2; deoxynucleotidyltransferase, terminal, interacting protein 2; DNTTIP2; ERBP; estrogen receptor binding protein; Estrogen receptor-binding protein; FCF2; HSU15552; LPTS-interacting protein 2; LPTS-RP2; TDIF2; tdT-interacting factor 2; Terminal deoxynucleotidyltransferase-interacting factor 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 30836 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5QJE6 | |
| DNTTIP2 | |
| synthetic peptide directed towards the C terminal region of human TDIF2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction