missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDI Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | TDI |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
TDI Polyclonal specifically detects TDI in Human samples. It is validated for Western Blot.Specifications
| TDI | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 100038246 | |
| Synthetic peptides corresponding to TD1(TLX1 divergent) The peptide sequence was selected from the middle region of TD1. Peptide sequence PPHPLNKQKHHPPHPSQTQKDLVPRSPQLEKSRIRLRRTLRNLGGGRGQR. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| APT-B7, Putative protein TDI, putative TLX1 neighbor protein, TD1, TDI, TLX1 divergent gene protein, TLX1 neighbor | |
| TLX1NB | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title