missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TDG Monoclonal Antibody (GT1511)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531522
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Human (99%), Mouse (88%), Rat (87%), Bovine (91%). Positive Control: human TDG-transfected 293T cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene belongs to the TDG/mug DNA glycosylase family. Thymine-DNA glycosylase removes thymine moieties from G/T mismatches by hydrolyzing the carbon-nitrogen bond between the sugar-phosphate backbone of DNA and the mis paired thymine. With lower activity, this enzyme also removes thymine from C/T and T/T mispairings. TDG can also remove uracil and 5-bromouracil from mispairings with guanine. This enzyme plays a central role in cellular defense against genetic mutation caused by the spontaneous deamination of 5-methylcytosine and cytosine. This gene may have a pseudogene in the p arm of chromosome 12.
Specifications
| TDG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P56581, Q13569 | |
| TDG | |
| Recombinant protein encompassing a sequence within the center region of human TDG. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| GT1511 | |
| Unconjugated | |
| TDG | |
| C-JUN leucine zipper interactive protein JZA-3; E130317C12Rik; G/T mismatch-specific thymine DNA glycosylase; hTDG; Jza1; JZA-3; Tdg; thymine DNA glycosylase; thymine-DNA glycosylase | |
| Mouse | |
| Protein A | |
| RUO | |
| 114521, 21665, 6996 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur