missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDG Antibody (1511), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-43718
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
TDG Monoclonal antibody specifically detects TDG in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin).
Specifications
| TDG | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| TDG | |
| Recombinant protein encompassing a sequence within the center region of human TDG. The exact sequence is proprietary. | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1511 | |
| Western Blot 1:5000-1:20000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| EC 3.2.2.29, thymine-DNA glycosylaseG/T mismatch-specific thymine DNA glycosylase | |
| Mouse | |
| 46 kDa | |
| 0.1 mL | |
| Base Excision Repair, DNA Repair | |
| 6996 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction