missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCTN3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598517
This item is not returnable.
View return policy
Description
TCTN3 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
TCTN3 is a member of the Tectonic protein family, a group of evolutionarily conserved secreted and transmembrane proteins that regulate the Hedgehog (Hh)-mediated patterning of the neural tube. While the expression pattern and function of TCTN3 is not known, it is 58% similar to TCTN1. TCTN1 is expressed during embryonic development in regions that participate in Hh signaling, beginning in the gastrulation stages in the ventral node. Mice expressing mutant TCTN1 die between E13.5 and E16.5 and display holoprosencephaly, a defect associated with reduced Hh signaling, indicating the role of TCTN1 as an Hh activator. At later stages in development, TCTN1 is thought to also act as a repressor on the Hh pathway in the anterior and posterior neural tube.
Specifications
| TCTN3 | |
| Polyclonal | |
| Unconjugated | |
| TCTN3 | |
| 4930521E07Rik; AI197391; C10orf61; JBTS18; OFD4; PSEC0041; RGD1305166; RP11-7D5.3; TCTN3; Tect3; tectonic 3; tectonic family member 3; tectonic-3; UNQ1881/PRO4324 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 26123, 309486 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 7 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q6NUS6 | |
| TCTN3 | |
| Recombinant Human Tectonic-3 protein (348-472AA). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction