missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCTN2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £364.00
Specifications
| Antigen | TCTN2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TCTN2 Polyclonal antibody specifically detects TCTN2 in Mouse samples. It is validated for Western BlotSpecifications
| TCTN2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 79867 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| C12orf38, chromosome 12 open reading frame 38, FLJ12975, TECT2tectonic 2, tectonic family member 2, tectonic-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 420-670 of human TCTN2 (NP_079085.2). VKFLSYNSGNEEELSGNPGYQLGKPVRALNINRMNNVTTLHLWQSAGRGLCTSATFKPILFGENVLSGCLLEVGINENCTQLRENAVERLDSLIQATHVAMRGNSDYADLSDGWLEIIRVDAPDPGADPLASSVNGMCLDIPAHLSIRILISDAGAVEGITQQEILGVETRFSSVNWQYQCGLTCEHKADLLPISASVQFIKIPAQLPHPLTRFQINYTEYDCNRNEVCWPQLLYPWTQYYQGELHSQCVA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title