missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCTN1 Polyclonal Antibody
Description
A suggested positive control is mouse kidney tissue lysate. PA5-20983 can be used with blocking peptide PEP-1097.
TCTN1 is the founding member of the Tectonic protein family, a group of evolutionarily conserved secreted and transmembrane proteins that regulate the Hedgehog (Hh)-mediated patterning of the neural tube. During embryonic development, TCTN1 is expressed in regions that participate in Hh signaling, beginning in the gastrulation stages in the ventral node. Mice expressing mutant TCTN1 die between E13. 5 and E16. 5 and display holoprosencephaly, a defect associated with reduced Hh signaling, indicating the role of TCTN1 as an Hh activator. At later stages in development, TCTN1 is thought to also act as a repressor on the Hh pathway in the anterior and posterior neural tube.
Specifications
Specifications
| Antigen | TCTN1 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | TCTN1 |
| Gene Accession No. | Q2MV58, Q8BZ64 |
| Gene Alias | G730031O11Rik; JBTS13; RGD1566266; TCTN1; TECT1; tectonic family member 1; tectonic-1; UNQ9369/PRO34160 |
| Gene Symbols | TCTN1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?