Learn More
Invitrogen™ TCTN1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA599977
Product Code. 16634225
  • £457.00 / 100µL
Description
Antibody detects endogenous levels of total TCTN1.
TCTN1 is the founding member of the Tectonic protein family, a group of evolutionarily conserved secreted and transmembrane proteins that regulate the Hedgehog (Hh)-mediated patterning of the neural tube. During embryonic development, TCTN1 is expressed in regions that participate in Hh signaling, beginning in the gastrulation stages in the ventral node. Mice expressing mutant TCTN1 die between E13. 5 and E16. 5 and display holoprosencephaly, a defect associated with reduced Hh signaling, indicating the role of TCTN1 as an Hh activator. At later stages in development, TCTN1 is thought to also act as a repressor on the Hh pathway in the anterior and posterior neural tube.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TCTN1
Polyclonal
Unconjugated
TCTN1
G730031O11Rik; JBTS13; RGD1566266; TCTN1; TECT1; tectonic family member 1; tectonic-1; UNQ9369/PRO34160
Rabbit
Affinity chromatography
RUO
304486, 654470, 79600
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q2MV58, Q8BZ64
TCTN1
A synthesized peptide derived from human TCTN1(Accession Q2MV58), corresponding to amino acid residues S37-C87.
100 μL
Primary
Human, Mouse, Rat